Ethanol, 96%, specified according to the requirements of Ph. Eur, denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex™
Thermo Scientific Chemicals

Ethanol, 96%, specified according to the requirements of Ph. Eur, denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex™

CAS: 64-17-5 | C2H6O | 46.07 g/mol
Have Questions?
Catalog NumberQuantity
4636900101 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 463690010
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS64-17-5
IUPAC Nameethanol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
View more
SpecificationsSpecification SheetSpecification Sheet
DenaturantDenatured with 1% iPr-OH, 1% MEK and 0.001% Bitrex
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use