Phosphoric acid, for HPLC
Thermo Scientific Chemicals

Phosphoric acid, for HPLC

CAS: 7664-38-2 | H3O4P | 97.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 463861000
Price (USD)
33.50
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
33.50
Each
Chemical Identifiers
CAS7664-38-2
IUPAC Namephosphoric acid
Molecular FormulaH3O4P
InChI KeyNBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILESOP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 210 nm A: =<0.07
UVat 230 nm A: =<0.04
UV(1 M solution)
UVat 250 nm A: =<0.02
Density1.70 kg/l (about)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only