DyLight™ 680 NHS Ester
Product Image
Thermo Scientific™

DyLight™ 680 NHS Ester

Thermo Scientific DyLight 680 Amine-Reactive Dye is an NHS ester-activated derivative of high-performance DyLight 680 used to fluorescently label antibodiesRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
464195 x 50 μg
464181 mg
Catalog number 46419
Price (KRW)
397,000
Online offer
Ends: 31-Mar-2026
466,000
Save 69,000 (15%)
Each
Add to cart
Quantity:
5 x 50 μg
Price (KRW)
397,000
Online offer
Ends: 31-Mar-2026
466,000
Save 69,000 (15%)
Each
Add to cart
Thermo Scientific DyLight 680 Amine-Reactive Dye is an NHS ester-activated derivative of high-performance DyLight 680 used to fluorescently label antibodies and other proteins that are then used as molecular probes for cellular imaging and other fluorescence detection methods.

DyLight 680 produces near-infrared (IR) fluorescence that replaces other near-IR dyes, including Cy5.5™ dye and Alexa Fluor™ 680, and is ideal for multi-color applications. The high water solubility of DyLight Fluors means that a high dye-to-protein ratio can be attained without causing precipitation of the conjugates. DyLight 680 Amine-Reactive Dye is also available as part of two antibody labeling kit sizes.

Features of DyLight 680 NHS-Ester:

High performance—DyLight 680 fluoresces brighter than Alexa Fluor 680 and Cy5.5 dye
Specific—NHS ester-activated dye labels proteins and other molecules at primary amines (-NH2)
Optimized procedure—following the standard protocol results in antibodies with excellent dye:protein ratios and recovery rates for optimum activity and fluorescence labeling

Applications:
• Primary antibody labeling for immunofluorescence microscopy, immunohistochemistry (IHC), Western blotting, or ELISA assay
• Target protein labeling for in vitro and in vivo fluorescent detection strategies

DyLight 680 Amine-Reactive Dye is activated with an N-hydroxysuccinimide (NHS) ester moiety to react with exposed N-terminal α-amino groups or the ε-amino groups of lysine residues to form stable amide bonds. Learn more about NHS ester chemistry.

Typical labeling reactions require the dye to first be dissolved in anhydrous dimethyl formamide (DMF) or another suitable organic solvent before adding a specific molar amount of dye to an amine-free buffer containing the protein to be labeled. However, the high solubility of DyLight Fluors permits protein solutions to be added directly to specific amounts of the labeling reagent. This feature allows DyLight 680 Amine-Reactive Dye to be provided in multiple formats with flexible protocols to achieve efficient degrees of labeling.

We also offer Standard and Microscale DyLight 680 Antibody Labeling Kits for fast and efficient fluorescent labeling of antibodies for use in fluorescence methods.The standard size kit contains all necessary components to perform three separate labeling reactions using 1 mg of IgG or similar quantities of other proteins. The microscale kit contains all of the necessary components to perform five separate labeling reactions using 100 μg of IgG. Both kit sizes include the Amine-Reactive DyLight 680 NHS-ester in convenient single-use vials as well as purification resin and spin columns for the preparation of ready-to-use conjugate.

Related Products
DyLight™ 680 Antibody Labeling Kit
DyLight™ 680 Microscale Antibody Labeling Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Emission712
Excitation692
Labeling TargetProteins
Label or DyeDyLight™ 680
Product TypeDye
Quantity5 x 50 μg
Reactive MoietyActive Ester, Succinimidyl Ester, NHS Ester
SolubilityDMF (Dimethylformamide)
Label TypeDyLight Fluors
Product LineDyLight
Unit SizeEach
Contents & Storage
Store desiccated at -20°C.