Product Image
Thermo Scientific Chemicals

Thiacloprid

CAS: 111988-49-9 | C10H9ClN4S | 252.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 464390010
Price (USD)
173.65
Special offer
Online exclusive
Ends: 28-Dec-2026
204.00
Save 30.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
173.65
Special offer
Online exclusive
Ends: 28-Dec-2026
204.00
Save 30.35
Each
Chemical Identifiers
CAS111988-49-9
IUPAC Name({3-[(6-chloropyridin-3-yl)methyl]-1,3-thiazolidin-2-ylidene}amino)formonitrile
Molecular FormulaC10H9ClN4S
InChI KeyHOKKPVIRMVDYPB-UHFFFAOYSA-N
SMILESClC1=CC=C(CN2CCSC2=NC#N)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point133°C to 138°C
HPLC>=95.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only