Product Image
Thermo Scientific Chemicals

Apixaban

CAS: 503612-47-3 | C25H25N5O4 | 459.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number 464495000
Price (USD)
-
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS503612-47-3
IUPAC Name1-(4-methoxyphenyl)-7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-1H,4H,5H,6H,7H-pyrazolo[3,4-c]pyridine-3-carboxamide
Molecular FormulaC25H25N5O4
InChI KeyQNZCBYKSOIHPEH-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)N1N=C(C(N)=O)C2=C1C(=O)N(CC2)C1=CC=C(C=C1)N1CCCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Melting point238°C to 240°C
HPLC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only