Product Image
Thermo Scientific Chemicals

Neratinib

CAS: 698387-09-6 | C30H29ClN6O3 | 557.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 464702500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS698387-09-6
IUPAC NameN-[4-({3-chloro-4-[(pyridin-2-yl)methoxy]phenyl}amino)-3-cyano-7-ethoxyquinolin-6-yl]-4-(dimethylamino)but-2-enamide
Molecular FormulaC30H29ClN6O3
InChI KeyJWNPDZNEKVCWMY-UHFFFAOYSA-N
SMILESCCOC1=C(NC(=O)C=CCN(C)C)C=C2C(NC3=CC(Cl)=C(OCC4=CC=CC=N4)C=C3)=C(C=NC2=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.5 %
Solubility(1% in DMSO) Clear colorless to light yellow
Appearance (Color)White to light yellow to beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only