Magnesium gluconate hydrate
Magnesium gluconate hydrate
Magnesium gluconate hydrate
Thermo Scientific Chemicals

Magnesium gluconate hydrate

CAS: 336879-53-9 | C12H24MgO15 | 432.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 464831000
Price (USD)
88.65
Special offer
Online exclusive
Ends: 28-Dec-2026
104.00
Save 15.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 28-Dec-2026
104.00
Save 15.35
Each
Chemical Identifiers
CAS336879-53-9
IUPAC Namemagnesium(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) hydrate
Molecular FormulaC12H24MgO15
InChI KeyIAKLPCRFBAZVRW-XRDLMGPZSA-L
SMILESO.[Mg++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration Complexometric98.0 to 102.0 % (on dry substance)
Water=<13.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only