Betamethasone valerate
Thermo Scientific Chemicals

Betamethasone valerate

製品番号(カタログ番号)数量
4649300505 g
4649300101 g
製品番号(カタログ番号) 464930050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2152-44-5
IUPAC Name(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-10-hydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl pentanoate
Molecular FormulaC27H37FO6
InChI KeySNHRLVCMMWUAJD-SUYDQAKGSA-N
SMILESCCCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO
さらに表示
HPLC>=97.5 %
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<0.5 % (1 g, 105°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only