Product Image
Thermo Scientific Chemicals

Elacridar

CAS: 143664-11-3 | C34H33N3O5 | 563.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 464950010
Price (USD)
1,587.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,868.00
Save 280.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
1,587.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,868.00
Save 280.35
Each
Chemical Identifiers
CAS143664-11-3
IUPAC NameN-{4-[2-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)ethyl]phenyl}-5-methoxy-9-oxo-9,10-dihydroacridine-4-carboxamide
Molecular FormulaC34H33N3O5
InChI KeyOSFCMRGOZNQUSW-UHFFFAOYSA-N
SMILESCOC1=C2NC3=C(C=CC=C3C(=O)C2=CC=C1)C(=O)NC1=CC=C(CCN2CCC3=CC(OC)=C(OC)C=C3C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Almost white to yellow
Appearance (Form)Powder
HPLC>=97.5 %
Water=<1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only