Product Image
Thermo Scientific Chemicals

Faropenem sodium hydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 464960010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS122547-49-3
IUPAC Namesodium (5R,6S)-6-[(1R)-1-hydroxyethyl]-7-oxo-3-[(2R)-oxolan-2-yl]-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate
Molecular FormulaC12H14NNaO5S
InChI KeyICSAXRANXQSPQP-VUKDEKJYSA-M
SMILES[Na+].[H][C@]12SC(=C(N1C(=O)[C@]2([H])[C@@H](C)O)C([O-])=O)[C@@]1([H])CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Form)Crystalline powder
Water12.0 to 13.5 %
Appearance (Color)White to light yellow to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only