Azasetron hydrochloride
Azasetron hydrochloride
Azasetron hydrochloride
Azasetron hydrochloride
Thermo Scientific Chemicals

Azasetron hydrochloride

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465070010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS123040-16-4
IUPAC Namehydrogen N-{1-azabicyclo[2.2.2]octan-3-yl}-6-chloro-4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxamide chloride
Molecular FormulaC17H21Cl2N3O3
InChI KeyDBMKBKPJYAHLQP-UHFFFAOYNA-N
SMILES[H+].[Cl-].CN1C(=O)COC2=C1C=C(Cl)C=C2C(=O)NC1CN2CCC1CC2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point301.0°C to 303.0°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only