Product Image
Thermo Scientific Chemicals

Azelastine hydrochloride

CAS: 79307-93-0 | C22H25Cl2N3O | 418.36 g/mol
Have Questions?
Catalog NumberQuantity
465082500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465082500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS79307-93-0
IUPAC Namehydrogen 4-[(4-chlorophenyl)methyl]-2-(1-methylazepan-4-yl)-1,2-dihydrophthalazin-1-one chloride
Molecular FormulaC22H25Cl2N3O
InChI KeyYEJAJYAHJQIWNU-UHFFFAOYNA-N
SMILES[H+].[Cl-].CN1CCCC(CC1)N1N=C(CC2=CC=C(Cl)C=C2)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only