Product Image
Thermo Scientific Chemicals

1-Isoindolinone

CAS: 480-91-1 | C8H7NO | 133.15 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 465180050
價格 (TWD)
-
Quantity:
5 g
Request bulk or custom format
化學識別
CAS480-91-1
IUPAC Name2,3-dihydro-1H-isoindol-1-one
Molecular FormulaC8H7NO
InChI KeyPXZQEOJJUGGUIB-UHFFFAOYSA-N
SMILESO=C1NCC2=CC=CC=C12
檢視更多
規格Specification Sheet規格表
Appearance (Form)Crystalline powder
Melting point149°C to 153°C
Infrared spectrumConforms
Appearance (Color)White to light yellow
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only