Product Image
Thermo Scientific Chemicals

Eriodictyol, 94+%

CAS: 552-58-9 | C15H12O6 | 288.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465242500
Price (USD)
191.00
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
191.00
Each
Chemical Identifiers
CAS552-58-9
IUPAC Name(2S)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-1-benzopyran-4-one
Molecular FormulaC15H12O6
InChI KeySBHXYTNGIZCORC-ZDUSSCGKSA-N
SMILESOC1=CC(O)=C2C(=O)C[C@H](OC2=C1)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only