Product Image
Thermo Scientific Chemicals

1-Methylimidazole, for biochemistry, anhydrous, AcroSeal™

Have Questions?
Catalog NumberQuantity
4652900101 L
Catalog number 465290010
Price (USD)
343.65
Special offer
Online exclusive
Ends: 28-Dec-2026
404.00
Save 60.35
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
343.65
Special offer
Online exclusive
Ends: 28-Dec-2026
404.00
Save 60.35
Each
Chemical Identifiers
CAS616-47-7
IUPAC Name1-methyl-1H-imidazole
Molecular FormulaC4H6N2
InChI KeyMCTWTZJPVLRJOU-UHFFFAOYSA-N
SMILESCN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
AppearanceMay discolor to yellow on storage
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only