Product Image
Thermo Scientific Chemicals

Parecoxib sodium salt

Have Questions?
Catalog NumberQuantity
4653800505 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465380050
Price (EUR)
694,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
694,00
Each
Chemical Identifiers
CAS198470-85-8
IUPAC Namesodium [4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzenesulfonyl](propanoyl)azanide
Molecular FormulaC19H17N2NaO4S
InChI KeyHQPVVKXJNZEAFW-UHFFFAOYSA-M
SMILES[Na+].CCC(=O)[N-]S(=O)(=O)C1=CC=C(C=C1)C1=C(C)ON=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder
HPLC>=98.5 %
Water=<3 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only