Product Image
Thermo Scientific Chemicals

Almotriptan malate

 
Have Questions?
Catalog NumberQuantity
465402500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465402500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS181183-52-8
IUPAC Name2-hydroxybutanedioic acid; dimethyl(2-{5-[(pyrrolidine-1-sulfonyl)methyl]-1H-indol-3-yl}ethyl)amine
Molecular FormulaC21H31N3O7S
InChI KeyQHATUKWEVNMHRY-UHFFFAOYNA-N
SMILESOC(CC(O)=O)C(O)=O.CN(C)CCC1=CNC2=CC=C(CS(=O)(=O)N3CCCC3)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to beige to light yellow
Appearance (Color)to light orange
Appearance (Form)Powder
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only