Product Image
Thermo Scientific Chemicals

Dexmedetomidine hydrochloride

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465430010
Price (USD)
616.65
Special offer
Online exclusive
Ends: 28-Dec-2026
725.00
Save 108.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
616.65
Special offer
Online exclusive
Ends: 28-Dec-2026
725.00
Save 108.35
Each
Chemical Identifiers
CAS145108-58-3
IUPAC Namehydrogen 5-[(1S)-1-(2,3-dimethylphenyl)ethyl]-1H-imidazole chloride
Molecular FormulaC13H17ClN2
InChI KeyVPNGEIHDPSLNMU-MERQFXBCSA-N
SMILES[H+].[Cl-].C[C@H](C1=CN=CN1)C1=CC=CC(C)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white to beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point153°C to 160°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only