Chlorpropham
Thermo Scientific Chemicals

Chlorpropham

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465580050
Price (EUR)
26,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
26,20
Each
Chemical Identifiers
CAS101-21-3
IUPAC Namepropan-2-yl N-(3-chlorophenyl)carbamate
Molecular FormulaC10H12ClNO2
InChI KeyCWJSHJJYOPWUGX-UHFFFAOYSA-N
SMILESCC(C)OC(=O)NC1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Melting point38°C to 43°C
Appearance (Form)Powder or crystals
GC>=97.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only