Product Image
Thermo Scientific Chemicals

Sorafenib tosylate

CAS: 475207-59-1 | C28H24ClF3N4O6S | 637.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465710010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS475207-59-1
IUPAC Name4-[4-({[4-chloro-3-(trifluoromethyl)phenyl]carbamoyl}amino)phenoxy]-N-methylpyridine-2-carboxamide; 4-methylbenzene-1-sulfonic acid
Molecular FormulaC28H24ClF3N4O6S
InChI KeyIVDHYUQIDRJSTI-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(O)(=O)=O.CNC(=O)C1=CC(OC2=CC=C(NC(=O)NC3=CC=C(Cl)C(=C3)C(F)(F)F)C=C2)=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Almost white to light brown
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only