Dypnone, Thermo Scientific Chemicals
Dypnone, Thermo Scientific Chemicals
Dypnone, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dypnone, Thermo Scientific Chemicals

Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 465850250
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
規格
Alpha VectorDYPNONE
CAS495-45-4
Chemical Name or MaterialDypnone
ColorColorless to Yellow
Formula Weight222.29
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only