5-Formyl-2-furancarboxylic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Formyl-2-furancarboxylic acid, Thermo Scientific Chemicals

製品番号(カタログ番号) 466000010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13529-17-4
IUPAC Name5-formylfuran-2-carboxylate
Molecular FormulaC6H3O4
InChI KeySHNRXUWGUKDPMA-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=C(O1)C=O
さらに表示
Titration with NaOH98.0 to 102.0 %
Melting point203°C to 212°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only