(-)-Arctiin
Thermo Scientific Chemicals

(-)-Arctiin

製品番号(カタログ番号) 466050010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS20362-31-6
IUPAC Name(3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[(3-methoxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}phenyl)methyl]oxolan-2-one
Molecular FormulaC27H34O11
InChI KeyXOJVHLIYNSOZOO-RVDKUWPISA-N
SMILESCOC1=CC=C(C[C@H]2COC(=O)[C@@H]2CC2=CC=C(OC3OC(CO)C(O)C(O)C3O)C(OC)=C2)C=C1OC
さらに表示
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point107°C to 111°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only