Thermo Scientific™

DyLight™ 415-Co1 NHS Ester

Catalog number:  46609

 Related applications: Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Label peptides, antibodies and other proteins at primary amines with the coumarin-based, 405 laser-detectable Thermo Scientific DyLight Blue-emitting Specialty Dye.

DyLight Blue-emitting Dye is a labeling agent that provides bright fluorescence detection for imaging and flow cytometry. Dyes that contain a greater number of negatively charged sulfonates generally will have greater water solubility than dyes with fewer sulfonates. More hydrophobic dyes often provide better cell penetration in vivo, while more hydrophilic dyes have less nonspecific binding potential. This dye contains 1 sulfonate for increased solubility and an amine-reactive NHS ester for simple modification of antibodies, proteins, peptides, or other biomolecules through amide bond formation.

Features of DyLight 415-Co1 NHS Ester:

NHS ester reactive group—allows immediate labeling of antibodies, proteins, peptides and other amine-containing molecules through amide bond formation
Water solubility—1 sulfonate group increases the solubility of this dye over over nonsulfonated dyes

Applications:
• Molecular imaging
• Microscopy
• Flow cytometry
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: DyLight™ Fluor (Specialty)
Chemical Reactivity: Amine
Excitation⁄Emission (nm): 418⁄463
Product Line: DyLight™
Product Size: 1 mg
Solubility: DMF (Dimethylformamide)

Contents & storage

Store desiccated at -20°C.

Documents

Manuals & protocols

Recommended products