Search
| Catalog Number | Volume (Metric) | Number of Channels |
|---|---|---|
| 4661080 | 1 to 10 μL | 16 |
| 4661090 | 5 to 50 μL | 16 |
Reduce the force needed to eject multiple tips with our innovative soft-touch tip ejection, one of many ergonomic features of theThermo Scientific™ Finnpipette™ F1 Multichannel Pipette. As with the single-channel Finnpipette F1 pipettes, the AVG mechanism ensures a high level of accuracy and precision. In addition, in the low volume models the super blow-out function ensures accurate dispensing for even the lowest volumes.