Product Image
Thermo Scientific Chemicals

Glyphosate

CAS: 1071-83-6 | C3H8NO5P | 169.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 466180050
Price (EUR)
186,65
Online exclusive
207,00
Save 20,35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
186,65
Online exclusive
207,00
Save 20,35 (10%)
Each
Chemical Identifiers
CAS1071-83-6
IUPAC Name2-[(phosphonomethyl)amino]acetic acid
Molecular FormulaC3H8NO5P
InChI KeyXDDAORKBJWWYJS-UHFFFAOYSA-N
SMILESOC(=O)CNCP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Appearance (Color)White to light yellow
Infrared spectrumConforms
HPLC>=94 %
Water=<1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only