Zoledronic acid monohydrate
Zoledronic acid monohydrate
Zoledronic acid monohydrate
Thermo Scientific Chemicals

Zoledronic acid monohydrate

CAS: 165800-06-6 | C5H12N2O8P2, C5 H10 N2 O7 P2 . H2 O | 290.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 466210050
Price (EUR)
142,65
Online exclusive
158,00
Save 15,35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
142,65
Online exclusive
158,00
Save 15,35 (10%)
Each
Chemical Identifiers
CAS165800-06-6
IUPAC Name[1-hydroxy-2-(1H-imidazol-1-yl)-1-phosphonoethyl]phosphonic acid hydrate
Molecular FormulaC5H12N2O8P2
InChI KeyFUXFIVRTGHOMSO-UHFFFAOYSA-N
SMILESO.OC(CN1C=CN=C1)(P(O)(O)=O)P(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying5 % to 8 %
Titration with NaOH97.5 to 102.5 % (dry substance)
pH2.0 to 4.0
Appearance (Color)White to beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only