Dihydralazine sulfate
Thermo Scientific Chemicals

Dihydralazine sulfate

Have Questions?
Catalog NumberQuantity
466231000100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466231000
Price (USD)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS7327-87-9
IUPAC Name1,4-dihydrazinylphthalazine; sulfuric acid
Molecular FormulaC8H12N6O4S
InChI KeyBWHAMWGGORIDBK-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.NNC1=C2C=CC=CC2=C(NN)N=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Loss on drying13 % to 15 % (1 g, 50°C, 5 h) (vacuum)
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only