Carisoprodol
Thermo Scientific Chemicals

Carisoprodol

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466240250
Price (EUR)
451,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
451,00
Each
Chemical Identifiers
CAS78-44-4
IUPAC Name2-[(carbamoyloxy)methyl]-2-methylpentyl N-(propan-2-yl)carbamate
Molecular FormulaC12H24N2O4
InChI KeyOFZCIYFFPZCNJE-UHFFFAOYNA-N
SMILESCCCC(C)(COC(N)=O)COC(=O)NC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Loss on drying=<0.5 % (1 g, 3 hr) (vacuum)
TLC>=97.5 %
Melting point92°C to 96°C
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only