Thermo Scientific™

DyLight™ 615-B1 NHS Ester

Catalog number:  46625

 Related applications: Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific DyLight 615-B1 NHS Ester is a benzopyrillium-based red-emitting specialty dye that can be used to label peptides, antibodies, and other proteins at primary amines. This dye has excitation and emission peaks at 623 and 643 nm, respectively (in ethanol) and an extinction coefficient of 200,000 M-1cm-1

General features of DyLight red-emitting specialty dyes:

Large selection—the largest family of dyes available for red-emitting fluorescence applications
NHS ester reactive group—allows immediate labeling of antibodies, proteins, peptides, and other amine-containing molecules through amide bond formation
Multiple solubility options—choose from hydrophilic to hydrophobic dyes to optimize the right dye label for the best performance in a given application

DyLight Red-emitting Dyes are a family of labeling agents that provide bright fluorescence detection for imaging. Dyes can be selected based upon their characteristic excitation and emission properties or relative hydrophilicity and hydrophobicity attributes. Dyes that contain a greater number of negatively charged sulfonates generally will have greater water solubility than dyes with fewer sulfonates. More hydrophobic dyes often provide better cell penetrating ability in vivo, while more hydrophilic dyes have less nonspecific binding potential. Each dye contains an amine-reactive NHS ester for simple modification of antibodies, proteins, peptides or other biomolecules through amide bond formation.

DyLight Red-emitting Dyes are a family of labeling agents that provide bright fluorescence detection for imaging. Dyes can be selected based upon their characteristic excitation and emission properties or relative hydrophilicity and hydrophobicity attributes. Dyes that contain a greater number of negatively charged sulfonates generally will have greater water solubility than dyes with fewer sulfonates. More hydrophobic dyes often provide better cell penetrating ability in vivo, while more hydrophilic dyes have less nonspecific binding potential. Each dye contains an amine-reactive NHS ester for simple modification of antibodies, proteins, peptides or other biomolecules through amide bond formation.

Criteria to consider when choosing a DyLight Red-emitting Specialty Dye
• Excitation and emission wavelengths—choose the best dye to match the excitation and emission capabilities of your instrument
• Water solubility—choose a dye based on its relative hydrophilicity, which directly correlates to the number of negatively-charged sulfonates it has on its core structure. More hydrophilic dyes are best at maintaining water solubility of a labeled antibody and limiting the nonspecific binding of the conjugate. More hydrophobic dyes often are best at penetrating tissues and cell membranes in vivo, meaning that dyes with fewer sulfonates may work best for some applications.
• DyLight dye selection—the broad selection of red-emitting dyes allows a number of candidate dyes to be tested in a given application for optimal performance.

Applications:
• Fluorescence imaging
• Confocal microscopy
• Flow cytometry
• Spectral fluorescence imaging
In vivo imaging
• Fluorescent western blotting
• Protein microarrays
• Antibody labeling
• Peptide labeling
• Fluorescence correlation spectroscopy
• Protein arrays
• Single molecule detection
• Nanoparticle conjugation
• Biotin/streptavidin conjugation

Related Products
DyLight™ 633-B1 NHS Ester
DyLight™ 633-B2 NHS Ester
DyLight™ 633-B3 NHS Ester
DyLight™ 635-B2 NHS Ester
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: DyLight™ Fluor (Specialty)
Chemical Reactivity: Amine
Excitation⁄Emission (nm): 623⁄643
Product Line: DyLight™
Product Size: 1 mg
Solubility: DMF (Dimethylformamide)

Contents & storage

Store desiccated at -20°C. Shipped at ambient temperature.

Documents

Manuals & protocols