Cyclobenzaprine hydrochloride
Thermo Scientific Chemicals

Cyclobenzaprine hydrochloride

CAS: 6202-23-9 | C20H21N·HCl, C20H22ClN | 311.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 466250050
Price (USD)
146.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
146.00
Each
Chemical Identifiers
CAS6202-23-9
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Infrared spectrumConforms
Loss on drying=<1 % (105°C)
Solubility(50 mg/ml in ethanol) clear colorless to light yellow
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only