DyLight™ 680-4xPEG Maleimide

Catalog number: 46626

Thermo Scientific™  Related applications: Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific DyLight 680-4xPEG Sulfhydryl-Reactive Dye is a maleimide-activated derivative of our high-performance DyLight 680 Dye used to fluorescently label cysteine-containing peptides, proteins or other biomolecular probes.

The DyLight 680-4xPEG Dye contains 4 polyethylene glycol (PEG) chains that are non-toxic, enhance fluorescence and reduce nonspecific binding of conjugates made with them. Conjugates made with DyLight 680-4xPEG Dye can be used as molecular probes for cellular imaging and other fluorescence detection methods. The far-red to NIR fluorescence properties of DyLight 680-4xPEG Dye make it especially useful in a variety of biological, chemical, and pharmaceutical applications, including in vivo imaging. The PEG chains also improve solubility of the dyes and labeled molecules in aqueous solution, aid in cell permeability and improve tissue retention.

Features of DyLight 680-4xPEG Maleimide:

High fluorescence intensity—significantly brighter fluorescence than Alexa Fluor™ 680
PEGylated—improves solubility in aqueous solution, aids in cell permeability and improves tissue retention in in vivo applications
Convenient kit sizes—standard and microscale kit sizes to match your experimental needs
Specific—maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)

Applications:
• Fluorescence microscopy
In vivo or ex vivo imaging
• Cell-based assays
• Flow cytometry/fluorescence-activated cell sorting (FACS)

DyLight 680-4xPEG Sulfhydryl-Reactive Dye is activated with a maleic acid imide (maleimide) moiety to form a reactive alkylation reagent. Labeling occurs through reaction of the maleimide-activated dye with reduced sulfhydryl groups (-SH) to form stable thioether bonds. Maleimides are specific for sulfhydryl groups between pH 6.5-7.5. Learn more about maleimide chemistry. To see fluorescence performance data for DyLight 680-4xPEG-labeled probes, see the page for the amine-reactive variety.

Related Products
DyLight™ 680-4xPEG Maleimide
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity: Thiol
Excitation⁄Emission (nm): 692⁄712
Label or Dye: DyLight™ 680
Product Line: DyLight™
Product Size: 1 mg
Solubility: DMF (Dimethylformamide)

Contents & storage

Store at < 4°C.

Documents

Manuals & protocols