Thermo Scientific™

DyLight™ 800-4xPEG Maleimide

Catalog number:  46627

 Related applications: Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific DyLight 800-4xPEG Sulfhydryl-Reactive Dye is a maleimide-activated derivative of our high-performance DyLight 800 Dye used to fluorescently label cysteine-containing peptides, proteins or other biomolecular probes.

The DyLight 800-4xPEG dye contains 4 polyethylene glycol (PEG) chains that are non-cytotoxic, enhance fluorescence, and reduce nonspecific binding of conjugates made with them. Conjugates made with DyLight 800-4xPEG Dye can be used as molecular probes for cellular imaging and other fluorescence detection methods. The NIR fluorescence properties of DyLight 800-4xPEG Dye make it especially useful in a variety of biological, chemical, and pharmaceutical applications, including in vivo imaging. The PEG chains also improve solubility of the dyes and labeled molecules in aqueous solution, aid in cell permeability and improve tissue retention.

Features of DyLight 800-4xPEG Maleimide:

High fluorescence intensity—fluorescence comparable to Alexa Fluor™ 800 and IRDye™ 800
PEGylated—improves solubility in aqueous solution and aids in cell permeability
Specific—maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)

Applications:
• Fluorescence microscopy
In vivo or ex vivo imaging
• Cell-based assays
• Flow cytometry/fluorescence-activated cell sorting (FACS)

DyLight 800-4xPEG Sulfhydryl-Reactive Dye is activated with a maleic acid imide (maleimide) moiety to form a reactive alkylation reagent. Labeling occurs through reaction of the maleimide-activated dye with reduced sulfhydryl groups (-SH) to form stable thioether bonds. Maleimides are specific for sulfhydryl groups between pH 6.5-7.5. Learn more about maleimide chemistry.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity: Thiol
Excitation⁄Emission (nm): 777⁄794
Label or Dye: DyLight™ 800
Product Line: DyLight™
Product Size: 1 mg
Solubility: DMF (Dimethylformamide)

Contents & storage

Store desiccated at -20°C.

Documents

Manuals & protocols

Recommended products