Triethylene glycol dimethacrylate, contains MEHQ as inhibitor
Thermo Scientific Chemicals

Triethylene glycol dimethacrylate, contains MEHQ as inhibitor

CAS: 109-16-0 | C14H22O6 | 286.32 g/mol
製品番号(カタログ番号) 466272500
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS109-16-0
IUPAC Name2-(2-{2-[(2-methylprop-2-enoyl)oxy]ethoxy}ethoxy)ethyl 2-methylprop-2-enoate
Molecular FormulaC14H22O6
InChI KeyHWSSEYVMGDIFMH-UHFFFAOYSA-N
SMILESCC(=C)C(=O)OCCOCCOCCOC(=O)C(C)=C
さらに表示
Stabilizer75 to 225 ppm Hydroquinone monomethylether (MEHQ)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only