Glycopyrronium bromide
Thermo Scientific Chemicals

Glycopyrronium bromide

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466320010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS596-51-0
IUPAC Name3-[(2-cyclopentyl-2-hydroxy-2-phenylacetyl)oxy]-1,1-dimethylpyrrolidin-1-ium bromide
Molecular FormulaC19H28BrNO3
InChI KeyVPNYRYCIDCJBOM-UHFFFAOYNA-M
SMILES[Br-].C[N+]1(C)CCC(C1)OC(=O)C(O)(C1CCCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=95.0 %
Titration Argentometric>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only