Argatroban
Thermo Scientific Chemicals

Argatroban

製品番号(カタログ番号) 466390010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS74863-84-6
IUPAC Name(2R,4R)-1-{5-[(diaminomethylidene)amino]-2-(3-methyl-1,2,3,4-tetrahydroquinoline-8-sulfonamido)pentanoyl}-4-methylpiperidine-2-carboxylic acid
Molecular FormulaC23H36N6O5S
InChI KeyKXNPVXPOPUZYGB-VSVYTNTFSA-N
SMILESC[C@@H]1CCN([C@H](C1)C(O)=O)C(=O)C(CCCN=C(N)N)NS(=O)(=O)C1=CC=CC2=C1NCC(C)C2
さらに表示
Water=<4 %
Appearance (Form)Powder
HPLC>=97.5 %
Appearance (Color)White to almost white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only