SuperSignal™ Western Blot Enhancer
Thermo Scientific™

SuperSignal™ Western Blot Enhancer

Thermo Scientific SuperSignal Western Blot Enhancer contains a membrane treatment reagent and a primary antibody diluent that increase both signalLeia mais
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
46640500 mL
4664150 mL
Número do catálogo 46640
Preço (BRL)
-
Quantity:
500 mL
Request bulk or custom format
Thermo Scientific SuperSignal Western Blot Enhancer contains a membrane treatment reagent and a primary antibody diluent that increase both signal intensity and sensitivity 3- to 10-fold compared to a detection performed without it.

Features of SuperSignal Western Blot Enhancer:

• Increase sensitivity—achieve 3- to 10-fold increase in signal intensity and sensitivity
• Improve specificity—improves signal-to-noise ratio for poor quality and low affinity antibodies
• Better clarity—reduces background for cleaner Western blots
• Membrane compatibility—provides effective signal enhancement with PVDF and nitrocellulose membranes
• Substrate compatibility—validated for use with chromogenic, chemiluminescent and fluorescent detection methods

When a protein or antigen is difficult to detect because of low abundance or poor immunoreactivity, use of SuperSignal Western Blot Enhancer can significantly reduce background and enhance detection of low-abundance and weakly immunoreactive antigens.
For Research Use Only. Not for use in diagnostic procedures.
Especificações
Detection MethodChemiluminescence, Fluorescence, Colorimetric
For Use With (Application)Western Blot
FormLiquid
Membrane CompatibilityNitrocellulose, PVDF
Product LineSuperSignal
Product TypeWestern Blot Enhancer
Quantity500 mL
Unit SizeEach
Conteúdo e armazenamento
Upon receipt store solutions at 4°C Products are shipped at ambient temperature.

Frequently asked questions (FAQs)

How can I reduce background with SuperSignal West Atto Ultimate Sensitivity Substrate?

Here are potential causes for high background and corresponding solutions:
- Too much HRP in the system: Further dilute the HRP-conjugate, see guidelines in Table 1 in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018858_SuperSignalWestAtto_UG.pdf).
- Inadequate blocking: Optimize blocking conditions.
- Inappropriate blocking reagent: Try a different blocking reagent. Use clear milk or StartingBlock blocking buffer instead of dry milk.
- Inadequate washing: Increase length, number or volume of washes.
- Film has been overexposed: Decrease exposure time or use Pierce Background Eliminator (Cat No. 21065).
- Concentration of antigen or antibody is too high: Decrease amount of antigen or antibody.
- Poor antibody specificity: Use SuperSignal Western Blot Enhancer (Cat. No. 46640).
We also recommend minimizing handling of the membrane, using clean gloves while handling, clean plasticware, glassware, forceps, or other contact surfaces. We also recommend trimming of the membrane to remove excess area.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Do I have to use a SuperSignal Western Blot substrate after using the SuperSignal Western Blot Enhancer?

Not necessarily. The SuperSignal Western Blot Enhancer will work with any chemiluminescent substrate (including SuperSignal Western Blot substrates), as well as colorimetric or fluorometric detection systems.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What is the difference between Western Blot Signal Enhancer and SuperSignal Western Blot Enhancer? Is there any benefit to one versus the other?

The reagents in both kits are different. Western Blot Signal Enhancer (Cat. No. 21050) is a 2-step pre-treatment that occurs before blocking. It basically relaxes the protein on the membrane making the epitope easier to detect. SuperSignal Western Blot Enhancer (Cat. Nos. 46640, 46641) contains a membrane treatment reagent and a primary antibody diluent that increase both signal intensity and sensitivity 3- to 10-fold compared to a detecion performed without it. We have found that SuperSignal Western Blot Enhancer shows much less background while enhancing signal and sensitivity when compared to Western Blot Signal Enhancer.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

When should I use the SuperSignal Western Blot Enhancer (Cat. Nos. 46640 and 46641)?

The SuperSignal Western Blot Enhancer kit contains two reagents:

- Antigen Pretreatment Solution: used after transferring proteins to the membrane and before blocking or staining the membrane
- The Primary Antibody Diluent: used to dilute the primary antibody

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.