Melanin, synthetic
Thermo Scientific Chemicals

Melanin, synthetic

CAS: 8049-97-6 | C18H10N2O4, C18H10N2O4 | 318.29 g/mol
数量:
250 mg
1 g
5 g
製品番号(カタログ番号) 466432500
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS8049-97-6
IUPAC Name6,14-dimethyl-4,12-diazapentacyclo[8.6.1.1²,⁵.0¹³,¹⁷.0⁹,¹⁸]octadeca-1(17),2,5,9(18),10,13-hexaene-7,8,15,16-tetrone
Molecular FormulaC18H10N2O4
InChI KeyXUMBMVFBXHLACL-UHFFFAOYSA-N
SMILESCC1=C2NC=C3C2=C(C2=CNC4=C(C)C(=O)C(=O)C3=C24)C(=O)C1=O
さらに表示
Appearance (Color)Brown to black
Appearance (Form)powder
Infrared spectrumConforms
Carbon (C)44 to 55 %
Solubility(20 mg/ml NH4OH 1N) Brown to very dark brown and black
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only