Tofacitinib citrate
Thermo Scientific Chemicals

Tofacitinib citrate

CAS: 540737-29-9 | C22H28N6O8, C16H20N6O·C6H8O7 | 504.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 466532500
Price (USD)
168.65
Online Exclusive
187.00
Save 18.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
168.65
Online Exclusive
187.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS540737-29-9
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid; 3-{4-methyl-3-[methyl({7H-pyrrolo[2,3-d]pyrimidin-4-yl})amino]piperidin-1-yl}-3-oxopropanenitrile
Molecular FormulaC22H28N6O8
InChI KeySYIKUFDOYJFGBQ-UHFFFAOYNA-N
SMILESOC(=O)CC(O)(CC(O)=O)C(O)=O.CC1CCN(CC1N(C)C1=C2C=CNC2=NC=N1)C(=O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Solubility(5 mg/ml DMSO,warmed) Clear solution
Appearance (Form)Powder
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only