Dorzolamide hydrochloride
Thermo Scientific Chemicals

Dorzolamide hydrochloride

製品番号(カタログ番号) 466612500
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS130693-82-2
IUPAC Namehydrogen (2R,4R)-4-(ethylamino)-2-methyl-1,1-dioxo-2H,3H,4H-1λ⁶-thieno[2,3-b]thiopyran-6-sulfonamide chloride
Molecular FormulaC10H17ClN2O4S3
InChI KeyOSRUSFPMRGDLAG-CIRBGYJCSA-N
SMILES[H+].[Cl-].CCN[C@@H]1C[C@@H](C)S(=O)(=O)C2=C1C=C(S2)S(N)(=O)=O
さらに表示
Appearance (Color)White to almost white
Loss on drying=<0.5 % (1 g, 105°C)
HPLC>=98.0 %
Infrared spectrumConforms
Appearance (Form)crystalline powder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only