Genipin
Thermo Scientific Chemicals

Genipin

CAS: 6902-77-8 | C11H14O5, C11H14O5 | 226.23 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 mg
1 g
產品號碼 466642500
價格 (TWD)
-
Quantity:
250 mg
Request bulk or custom format
化學識別
CAS6902-77-8
IUPAC Namemethyl (1R,4aS,7aS)-1-hydroxy-7-(hydroxymethyl)-1H,4aH,5H,7aH-cyclopenta[c]pyran-4-carboxylate
Molecular FormulaC11H14O5
InChI KeyAZKVWQKMDGGDSV-BCMRRPTOSA-N
SMILESCOC(=O)C1=CO[C@@H](O)[C@H]2[C@@H]1CC=C2CO
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point118.0°C to 123.0°C
HPLC>=98.0 %
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only