Locust bean gum
Thermo Scientific Chemicals

Locust bean gum

CAS: 9000-40-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 466672500
Price (USD)
68.00
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
68.00
Each
Chemical Identifiers
CAS9000-40-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only