Idebenone
Thermo Scientific Chemicals

Idebenone

CAS: 58186-27-9 | C19H30O5, C19H30O5 | 338.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 466770010
Price (USD)
89.60
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
89.60
Each
Chemical Identifiers
CAS58186-27-9
IUPAC Name2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
Molecular FormulaC19H30O5
InChI KeyJGPMMRGNQUBGND-UHFFFAOYSA-N
SMILESCOC1=C(OC)C(=O)C(CCCCCCCCCCO)=C(C)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)powder
Melting point51.0°C to 55.0°C
Solubility(5% in Methanol) Clear to slight turbid,
Loss on drying=<0.5 %
Solubilityyellow to orange solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only