Mequitazine
Thermo Scientific Chemicals

Mequitazine

Have Questions?
Catalog NumberQuantity
4669300505 g
Catalog number 466930050
Price (USD)
321.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
321.00
Each
Chemical Identifiers
CAS29216-28-2
IUPAC Name10-({1-azabicyclo[2.2.2]octan-3-yl}methyl)-10H-phenothiazine
Molecular FormulaC20H22N2S
InChI KeyHOKDBMAJZXIPGC-UHFFFAOYNA-N
SMILESC(C1CN2CCC1CC2)N1C2=CC=CC=C2SC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)crystalline powder
HPLC>=98.0 %
Loss on drying=<1 % (1 g, 105°C)
Melting point146.0°C to 150.0°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only