Alosetron hydrochloride
Thermo Scientific Chemicals

Alosetron hydrochloride

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 466972500
Price (EUR)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS122852-69-1
IUPAC Name5-methyl-2-[(5-methyl-1H-imidazol-4-yl)methyl]-1H,2H,3H,4H,5H-pyrido[4,3-b]indol-1-one hydrochloride
Molecular FormulaC17H19ClN4O
InChI KeyFNYQZOVOVDSGJH-UHFFFAOYSA-N
SMILESCl.CN1C2=C(C3=CC=CC=C13)C(=O)N(CC1=C(C)NC=N1)CC2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder
Titration Argentometric97.5 to 102.5 %
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only