Immuno Tubes and Stoppers
Imagem ilustrativa. O produto pode variar
Immuno Tubes and Stoppers
Immuno Tubes and Stoppers
Thermo Scientific™
Immuno Tubes and Stoppers
Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoMaterialDiameter (Metric) OuterProduct TypeFor Use With (Equipment)
466982Polyethylene11 mmImmuno Tubes and Stopper
341866Polyethylene11 mmStopper11 mm Nunc-Immuno Tubes
343036Polyethylene15 mmStopper15 mm Nunc-Immuno Tubes
348801Polyethylene12 mmStopper12 mm Nunc-Immuno Tubes
443990Polyethylene12 mmImmuno Tubes and StopperImmuno Tube
444202Polystyrene12 mmImmuno Tubes and StopperImmuno Tube
468608Polyethylene15 mmImmuno Tubes and StopperImmuno Tube
470319Polystyrene12 mmImmuno Star Tubes and StopperImmuno Tube
Número do catálogo 466982
Preço (BRL)
-
Material:
Polyethylene
Diameter (Metric) Outer:
11 mm
Product Type:
Immuno Tubes and Stopper
Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.
Features:
  • Offered in choice of three surfaces for optimal binding
  • Three tube sizes with stoppers available (ordered separately)
  • Fits standard equipment
Choice of Surface:
  • Thermo Scientific™ PolySorp™ is a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains.
  • Thermo Scientific™ MaxiSorp™ is a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character.
  • Thermo Scientific™ Mini-Sorp™ is a polyethylene-based material that provides a low-binding surface.
Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):
  • MiniSorp surface
  • Molded from special polyethylene formulation
  • For liquid phase immunoasssays, including RIA, for storing of reagents, or collecting chromatographic fractions
Solid Phase Immunoassay Tubes:
  • PolySorp and MaxiSorp surfaces available for choice of optimal binding
  • For solid phase immuno techniques such as IRMA, ELISA and ILMA
  • High–quality polystyrene for optimal quality
  • StarTube design gives higher sensitivity and faster assay times
  • Enhances assay sensitivity and reduces incubation times
Recommended for:
  • ELISA
  • Solid Phase RIA
  • Liquid Phase RIA
  • Sample storage
Especificações
Diameter (Metric) Outer11 mm
No. per Case3600
No. per Pack150
Length (Metric)70 mm
MaterialPolyethylene
Product TypeImmuno Tubes and Stopper
SurfaceMiniSorp
Volume (Metric)4 mL
Unit SizeCase of 150