Tiaprida hidrocloruro
Thermo Scientific Chemicals

Tiaprida hidrocloruro

Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo 467200010
Precio (USD)
-
Cantidad:
1 g
Pedido a granel o personalizado
Identificadores químicos
CAS51012-33-0
IUPAC Namehydrogen N-[2-(diethylamino)ethyl]-5-methanesulfonyl-2-methoxybenzamide chloride
Molecular FormulaC15H25ClN2O4S
InChI KeyOTFDPNXIVHBTKW-UHFFFAOYSA-N
SMILES[H+].[Cl-].CCN(CC)CCNC(=O)C1=CC(=CC=C1OC)S(C)(=O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Loss on drying=<0.5 % (1 g, 105°C)
Titration with HClO498.5 to 101.5 %
HPLC>=98.5 %
Infrared spectrumConforms
Appearance (Color)White to almost white
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only