Potassium phosphate tribasic, 90+%, pure
Thermo Scientific Chemicals

Potassium phosphate tribasic, 90+%, pure

CAS: 7778-53-2 | K3O4P | 212.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Catalog number 467330250
Price (USD)
832.00
Each
Quantity:
25 kg
Request bulk or custom format
Price (USD)
832.00
Each
Chemical Identifiers
CAS7778-53-2
IUPAC Nametripotassium phosphate
Molecular FormulaK3O4P
InChI KeyLWIHDJKSTIGBAC-UHFFFAOYSA-K
SMILES[K+].[K+].[K+].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration excess HCl>=90.0 %
Water=<10 %
Appearance (Form)Powder or granules and/or chunks
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only