Xanthane hydride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Xanthane hydride, Thermo Scientific Chemicals

Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467340050
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6846-35-1
IUPAC Name5-amino-3H-1,2,4-dithiazole-3-thione
Molecular FormulaC2H2N2S3
InChI KeyYWZHEXZIISFIDA-UHFFFAOYSA-N
SMILESNC1=NC(=S)SS1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=94 %
Water=<1 %
Appearance (Color)Light yellow to brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only