Levocetirizine dihydrochloride
Thermo Scientific Chemicals

Levocetirizine dihydrochloride

Have Questions?
Catalog NumberQuantity
4674700101 g
Catalog number 467470010
Price (USD)
53.65
Online Exclusive
59.90
Save 6.25 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.90
Save 6.25 (10%)
Each
Chemical Identifiers
CAS130018-87-0
IUPAC Name2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dihydrochloride
Molecular FormulaC21H27Cl3N2O3
InChI KeyPGLIUCLTXOYQMV-UHFFFAOYNA-N
SMILESCl.Cl.OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point212°C to 216°C
Loss on drying=<0.5 %
Infrared spectrumConforms
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only